siRNA Details
Gene Symbol: Prnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012631.2 | NP_036763.1 | 3 | 605 | ||
XM_006235062.2 | XP_006235124.1 | 4 | 757 | |||
GenBank mRNA | BC072692.1 | - | 2 | 543 | 7099864 | |
FQ211780.1 | - | 685 | ||||
M20313.1 | - | 1 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
