siRNA Details
Gene Symbol: Ptpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012637.2 | NP_036769.1 | 8, 9 | 1161 | ||
XM_006235638.2 | XP_006235700.1 | 8, 9 | 1230 | |||
XM_006235639.2 | XP_006235701.1 | 8, 9 | 1225 | |||
GenBank mRNA | BC081829.1 | - | 8, 9 | 1161 | 7135855 | |
M33962.1 | - | 8, 9 | 1203 |
