siRNA Details
Gene Symbol: Raf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012639.2 | NP_036771.1 | 14 | 1746 | ||
XM_008763217.1 | XP_008761439.1 | 15 | 1876 | |||
XM_008763218.1 | XP_008761440.1 | 15 | 2056 | |||
GenBank mRNA | BC062071.1 | - | 14 | 1746 | 5599540 | |
M15427.1 | - | 1490 |
