siRNA Details
Gene Symbol: Rbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012640.2 | NP_036772.2 | 2 | 224 | ||
XM_006243607.1 | XP_006243669.1 | 3 | 404 | |||
GenBank mRNA | M13949.1 | - | 2 | 224 |

More Information
Other Species Targeted

Gene Ontology Categories:
