siRNA Details
Gene Symbol: RT1-EC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012645.1 | NP_036777.1 | 3 | 536 | ||
GenBank mRNA | AJ004887.1 | - | 2 | 455 | ||
AJ004888.1 | - | 2 | 455 | |||
AJ005024.1 | - | 2 | 455 | |||
AJ005025.1 | - | 2 | 455 | |||
BC061817.1 | - | 4 | 537 | 5598840 | ||
M31038.1 | - | 3 | 536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
