siRNA Details
Gene Symbol: Acsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033231.1 | NP_150234.1 | 3 | 477 | ||
XM_006230105.2 | XP_006230167.1 | 3 | 496 | |||
XM_006230106.2 | XP_006230168.1 | 3 | 457 | |||
GenBank mRNA | BC090325.1 | - | 3 | 477 | 7128525 |
