siRNA Details
Gene Symbol: Scnn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012648.1 | NP_036780.1 | 12 | 1539 | ||
XM_008759682.1 | XP_008757904.1 | 12 | 1574 | |||
XM_008759683.1 | XP_008757905.1 | 13 | 1784 | |||
XM_008759684.1 | XP_008757906.1 | 12 | 1616 | |||
XM_008759685.1 | XP_008757907.1 | 12 | 1593 | |||
XM_008759686.1 | XP_008757908.1 | 12 | 1593 | |||
GenBank mRNA | EU627516.1 | - | 11 | 1484 | ||
U35174.1 | - | 11 | 1494 | |||
U35175.1 | - | 11 | 1494 | |||
X77932.1 | - | 12 | 1556 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
