siRNA Details
Gene Symbol: Scnn1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017046.1 | NP_058742.1 | 3, 4 | 716 | ||
GenBank mRNA | EU627517.1 | - | 2, 3 | 618 | ||
U37539.1 | - | 3, 4 | 670 | |||
U37540.1 | - | 3, 4 | 670 | |||
X77933.1 | - | 3, 4 | 716 | |||
X78034.1 | - | 3, 4 | 699 |
