siRNA Details
Gene Symbol: Shbg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012650.1 | NP_036782.1 | 2 | 211 | ||
XM_006246586.2 | XP_006246648.1 | 3 | 960 | |||
XM_006246592.2 | XP_006246654.1 | 2 | 318 | |||
XM_008767771.1 | XP_008765993.1 | 3 | 425 | |||
XM_008767772.1 | XP_008765994.1 | 2 | 233 | |||
GenBank mRNA | BC097336.1 | - | 2 | 199 | 7453185 | |
M15034.1 | - | 2 | 211 | |||
M31179.1 | - | 2 | 163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
