siRNA Details
Gene Symbol: Slc10a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017047.1 | NP_058743.1 | 2 | 627 | ||
GenBank mRNA | BC091152.1 | - | 2 | 577 | 7370832 | |
FQ218559.1 | - | 2 | 589 | |||
M77479.1 | - | 2 | 627 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
