siRNA Details
Gene Symbol: Slc9a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173098.1 | NP_775121.1 | 9 | 2252 | ||
XM_006244738.2 | XP_006244800.1 | 9 | 2418 | |||
XM_008766988.1 | XP_008765210.1 | 9 | 2418 | |||
XM_008766989.1 | XP_008765211.1 | 10 | 2321 | |||
XM_008766990.1 | XP_008765212.1 | 10 | 2194 | |||
XM_008766991.1 | XP_008765213.1 | 9 | 1541 | |||
GenBank mRNA | M85301.1 | - | 9 | 2252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
