siRNA Details
Gene Symbol: Sod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017051.2 | NP_058747.1 | 2 | 258 | ||
GenBank mRNA | BC070913.1 | - | 2 | 258 | 7104698 | |
GQ404503.1 | - | 2 | 175 | |||
Y00497.1 | - | 2 | 186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
