siRNA Details
Gene Symbol: Sord | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017052.2 | NP_058748.2 | 3 | 318 | ||
GenBank mRNA | BC088398.1 | - | 3 | 318 | 7112546 | |
BC098919.1 | - | 3 | 265 | 7373210 | ||
BC128707.1 | - | 3 | 294 | 8373758 | ||
FQ209461.1 | - | 3 | 312 | |||
FQ210296.1 | - | 3 | 315 | |||
FQ211554.1 | - | 3 | 317 | |||
FQ234001.1 | - | 3 | 207 | |||
X59037.1 | - | 3 | 258 |
