siRNA Details
Gene Symbol: Tf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013110.1 | NP_001013128.1 | 13 | 1632 | ||
GenBank mRNA | AF476964.1 | - | 13 | 1663 | ||
AY327504.1 | - | 13 | 1601 | |||
BC087021.1 | - | 13 | 1632 | 7132382 | ||
D38380.1 | - | 13 | 1631 | |||
FQ209414.1 | - | 6 | 596 | |||
FQ209524.1 | - | 6 | 691 | |||
FQ209552.1 | - | 4 | 411 | |||
FQ209673.1 | - | 7 | 799 | |||
FQ209794.1 | - | 5 | 484 | |||
FQ210028.1 | - | 2 | 217 | |||
FQ210115.1 | - | 1 | 112 | |||
FQ210327.1 | - | 1 | 88 | |||
FQ210637.1 | - | 1 | 31 | |||
FQ210963.1 | - | 2 | 156 | |||
FQ211385.1 | - | 4 | 408 | |||
FQ213988.1 | - | 3 | 317 | |||
FQ218210.1 | - | 5 | 462 | |||
FQ219264.1 | - | 7 | 735 | |||
FQ219315.1 | - | 2 | 273 | |||
FQ219349.1 | - | 5 | 465 | |||
FQ219664.1 | - | 2 | 126 | |||
M27966.1 | - | 1 | 41 | |||
X77158.1 | - | 13 | 1609 |
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY325214.1 | - | 1599 | |||
AY325230.1 | - | 1602 | ||||
AY325241.1 | - | 1599 |
