siRNA Details
Gene Symbol: Tf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013110.1 | NP_001013128.1 | 9 | 1218 | ||
GenBank mRNA | AF476964.1 | - | 9 | 1249 | ||
AY327504.1 | - | 9 | 1187 | |||
BC087021.1 | - | 9 | 1218 | 7132382 | ||
D38380.1 | - | 9 | 1217 | |||
FQ209414.1 | - | 2 | 182 | |||
FQ209524.1 | - | 2 | 277 | |||
FQ209673.1 | - | 3 | 385 | |||
FQ209674.1 | - | 3 | 442 | |||
FQ209794.1 | - | 1 | 70 | |||
FQ214362.1 | - | 2 | 295 | |||
FQ218210.1 | - | 1 | 48 | |||
FQ219264.1 | - | 2 | 321 | |||
FQ219349.1 | - | 1 | 51 | |||
FQ219724.1 | - | 3 | 349 | |||
X77158.1 | - | 9 | 1195 |
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY325214.1 | - | 1185 | |||
AY325230.1 | - | 1185 | ||||
AY325241.1 | - | 1185 |
