siRNA Details
Gene Symbol: Tgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012671.2 | NP_036803.1 | 3 | 328 | ||
XM_006236822.2 | XP_006236884.1 | 3 | 1221 | |||
XM_008763049.1 | XP_008761271.1 | 3 | 1225 | |||
XM_008763050.1 | XP_008761272.1 | 3 | 1222 | |||
GenBank mRNA | KF366251.1 | - | 3 | 184 | ||
M31076.1 | - | 3 | 328 | |||
X02004.1 | - | 4 | 328 | |||
X61485.1 | - | 2 | 109 | |||
X61487.1 | - | 2 | 109 |

More Information
Other Species Targeted

Gene Ontology Categories:
