siRNA Details
Gene Symbol: Hist1h2ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022643.1 | NP_072169.1 | 1 | 120 | ||
GenBank mRNA | M18045.1 | - | 1 | 120 | ||
M18046.1 | - | 1 | 120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
