siRNA Details
Gene Symbol: Tpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012678.2 | NP_036810.1 | 5 | 506 | ||
GenBank mRNA | FQ225054.1 | - | 517 | |||
FQ225082.1 | - | 517 | ||||
J02780.1 | - | 5 | 509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
