siRNA Details
Gene Symbol: Clu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053021.2 | NP_444180.2 | 4 | 415 | ||
XM_006252094.2 | XP_006252156.1 | 3 | 293 | |||
GenBank mRNA | BC061534.1 | - | 3 | 194 | 6886725 | |
FQ209467.1 | - | 3 | 191 | |||
FQ209498.1 | - | 3 | 192 | |||
FQ209623.1 | - | 3 | 190 | |||
FQ211071.1 | - | 3 | 190 | |||
FQ213433.1 | - | 3 | 189 | |||
FQ214098.1 | - | 3 | 235 | |||
FQ218237.1 | - | 3 | 190 | |||
FQ218812.1 | - | 3 | 190 | |||
FQ219030.1 | - | 3 | 190 | |||
M16975.1 | - | 4 | 415 | |||
M64723.1 | - | 3 | 180 | |||
X13231.1 | - | 3 | 179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
