siRNA Details
Gene Symbol: Vhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052801.1 | NP_434688.1 | 3 | 595 | ||
GenBank mRNA | S80345.1 | - | 3 | 536 | ||
U14746.1 | - | 3 | 595 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
