siRNA Details
Gene Symbol: Vipr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012685.2 | NP_036817.1 | 4 | 930 | ||
GenBank mRNA | BC087136.1 | - | 4 | 930 | 7124910 | |
M86835.1 | - | 4 | 899 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
