siRNA Details
Gene Symbol: Yes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033298.1 | NP_150640.1 | 10 | 1481 | ||
XM_006245675.2 | XP_006245737.1 | 10 | 1494 | |||
GenBank mRNA | AB037472.1 | - | 9 | 1258 | ||
BC079403.1 | - | 10 | 1481 | 7112783 |
