siRNA Details
Gene Symbol: Abcb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012690.2 | NP_036822.1 | 21 | 2566 | ||
XM_006235999.2 | XP_006236061.1 | 19 | 2626 | |||
XM_006236000.1 | XP_006236062.1 | 17 | 2289 | |||
XM_008762697.1 | XP_008760919.1 | 20 | 2643 | |||
GenBank mRNA | L15079.1 | - | 21 | 2568 |
