siRNA Details
Gene Symbol: Sult2a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012695.3 | NP_036827.3 | 1 | 103 | ||
GenBank mRNA | M33329.1 | - | 1 | 103 | ||
X63410.1 | - | 1 | 103 |
Gene Symbol: Sult2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131903.1 | NP_571978.1 | 1 | 103 | ||
XM_008758891.1 | XP_008757113.1 | 1 | 133 | |||
XM_008758892.1 | XP_008757114.1 | 1 | 132 | |||
XM_008758893.1 | XP_008757115.1 | 1 | 121 | |||
GenBank mRNA | D14987.1 | - | 1 | 103 | ||
D14988.1 | - | 1 | 103 | |||
M31363.1 | - | 1 | 103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
