siRNA Details
Gene Symbol: Stx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012700.2 | NP_036832.1 | 7 | 529 | ||
XM_008759859.1 | XP_008758081.1 | 7 | 1066 | |||
XM_008759860.1 | XP_008758082.1 | 7 | 467 | |||
GenBank mRNA | M95735.1 | - | 7 | 688 |
