siRNA Details
Gene Symbol: Gcgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172091.2 | NP_742088.1 | 4 | 321 | ||
NM_172092.2 | NP_742089.1 | 4 | 329 | |||
XM_006247876.2 | XP_006247938.1 | 4 | 435 | |||
XM_006247877.1 | XP_006247939.1 | 2 | 165 | |||
XM_008768468.1 | XP_008766690.1 | 3 | 378 | |||
XM_008768469.1 | XP_008766691.1 | 3 | 381 | |||
GenBank mRNA | BC081700.1 | - | 4 | 293 | 7192550 | |
L04796.1 | - | 4 | 291 | |||
M96674.1 | - | 4 | 331 | |||
X68692.1 | - | 4 | 291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
