siRNA Details
Gene Symbol: Pgam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017328.2 | NP_059024.1 | 2 | 563 | ||
GenBank mRNA | FQ214696.1 | - | 2 | 576 | ||
FQ214983.1 | - | 2 | 576 | |||
FQ215677.1 | - | 2 | 576 | |||
FQ217291.1 | - | 2 | 583 | |||
FQ218013.1 | - | 2 | 576 | |||
FQ223775.1 | - | 2 | 576 | |||
FQ224280.1 | - | 2 | 576 | |||
FQ224543.1 | - | 2 | 579 | |||
M31835.1 | - | 2 | 564 |
