siRNA Details
Gene Symbol: RT1-A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008827.1 | NP_001008827.1 | 4, 5 | 906 | ||
XM_006256099.2 | XP_006256161.1 | 4, 5 | 937 | |||
GenBank mRNA | FQ228888.1 | - | 4, 5 | 953 | ||
FQ231496.1 | - | 4, 5 | 926 | |||
KC222922.1 | - | 3, 4 | 811 | |||
U50448.1 | - | 4, 5 | 897 | |||
X90375.1 | - | 3, 4 | 834 |
Gene Symbol: RT1-EC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ249700.1 | - | 4, 5 | 834 | ||
AJ249702.1 | - | 4, 5 | 834 | |||
U50447.1 | - | 6, 7 | 897 | |||
Y08532.1 | - | 4, 5 | 834 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
