siRNA Details
Gene Symbol: Tacr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080768.1 | NP_542946.1 | 1 | 659 | ||
GenBank mRNA | M31838.1 | - | 1 | 659 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
