siRNA Details
Gene Symbol: Snap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030991.3 | NP_112253.1 | 405 | |||
GenBank mRNA | AB003992.1 | - | 196 | |||
AF245227.1 | - | 404 | ||||
BC087699.1 | - | 360 | 7314318 | |||
FQ212040.1 | - | 376 | ||||
FQ213289.1 | - | 373 | ||||
FQ213923.1 | - | 376 | ||||
FQ214162.1 | - | 405 | ||||
U56262.1 | - | 169 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011428, M22012, AF483516, BC018249, AK078038, and others.
Gene Ontology Categories:
