siRNA Details
Gene Symbol: Itgam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012711.1 | NP_036843.1 | 17, 18 | 2281 | ||
XM_006230214.2 | XP_006230276.1 | 18, 19 | 2404 | |||
GenBank mRNA | AF268593.1 | - | 17, 18 | 2281 |
