siRNA Details
Gene Symbol: Adm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012715.1 | NP_036847.1 | 2 | 256 | ||
XM_008759687.1 | XP_008757909.1 | 3 | 1763 | |||
GenBank mRNA | BC061775.1 | - | 2 | 252 | 5621783 | |
D15069.1 | - | 2 | 238 | |||
U15419.1 | - | 2 | 256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
