siRNA Details
Gene Symbol: Slc16a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012716.2 | NP_036848.1 | 4 | 1021 | ||
GenBank mRNA | AJ236865.1 | - | 1 | 41 | ||
BC078877.1 | - | 4 | 1021 | 7122399 | ||
D63834.1 | - | 1035 | ||||
X86216.1 | - | 1041 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
