siRNA Details
Gene Symbol: Plcb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024353.1 | NP_077329.1 | 23 | 2158 | ||
GenBank mRNA | AF031370.1 | - | 21 | 1925 | ||
L15556.1 | - | 23 | 2158 | |||
L18962.1 | - | 23 | 2058 | |||
U57836.1 | - | 23 | 2058 |
