siRNA Details
Gene Symbol: Sstr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012719.2 | NP_036851.1 | 2 | 919 | ||
XM_006240115.2 | XP_006240177.1 | 2 | 2222 | |||
GenBank mRNA | L02915.1 | - | 2 | 693 | ||
M97656.1 | - | 1 | 577 | |||
X62314.1 | - | 1 | 601 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
