siRNA Details
Gene Symbol: Sds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053962.3 | NP_446414.3 | 3, 4 | 295 | ||
XM_008769213.1 | XP_008767435.1 | 2, 3 | 793 | |||
GenBank mRNA | BC088110.1 | - | 3, 4 | 295 | 7323391 | |
J03863.1 | - | 3, 4 | 385 | |||
Y00752.1 | - | 3, 4 | 312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
