siRNA Details
Gene Symbol: Ntrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163168.2 | NP_001156640.1 | 9 | 1544 | ||
NM_012731.2 | NP_036863.1 | 9 | 1529 | |||
XM_006253507.2 | XP_006253569.1 | 9 | 1480 | |||
XM_008771425.1 | XP_008769647.1 | 9 | 1485 | |||
XM_008771426.1 | XP_008769648.1 | 9 | 1371 | |||
XM_008771427.1 | XP_008769649.1 | 9 | 1479 | |||
GenBank mRNA | AY265419.1 | - | 9 | 1309 | ||
M55291.1 | - | 9 | 1532 | |||
M55292.1 | - | 9 | 1309 | |||
M55293.1 | - | 8 | 976 |
