siRNA Details
Gene Symbol: Ntrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012731.2 | NP_036863.1 | 15 | 2412 | ||
XM_008771425.1 | XP_008769647.1 | 16 | 2416 | |||
XM_008771426.1 | XP_008769648.1 | 16 | 2302 | |||
GenBank mRNA | M55291.1 | - | 15 | 2415 |
