siRNA Details
Gene Symbol: Rbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012733.4 | NP_036865.1 | 2 | 257 | ||
GenBank mRNA | FQ209770.1 | - | 2 | 271 | ||
FQ209927.1 | - | 2 | 241 | |||
FQ210835.1 | - | 2 | 272 | |||
FQ218090.1 | - | 2 | 271 | |||
M16459.1 | - | 2 | 257 | |||
M19257.1 | - | 2 | 187 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011254, BC018254, AK011641, BC091751, X60367.
Gene Ontology Categories:
