siRNA Details
Gene Symbol: Crabp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105716.1 | NP_001099186.1 | 1 | 153 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001284507, NM_013496, BC065787, X15789, X15481, and others.
Gene Ontology Categories:
