siRNA Details
Gene Symbol: Chrnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052806.2 | NP_434693.1 | 5 | 1007 | ||
GenBank mRNA | AY574260.1 | - | 5 | 1007 | ||
U42976.1 | - | 1067 | ||||
X15834.1 | - | 1113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
