siRNA Details
Gene Symbol: Avpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053019.2 | NP_444178.2 | 1 | 990 | ||
GenBank mRNA | BC088095.1 | - | 1 | 974 | 7321427 | |
Z11690.1 | - | 1 | 998 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Avpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053019.2 | NP_444178.2 | 1 | 990 | ||
GenBank mRNA | BC088095.1 | - | 1 | 974 | 7321427 | |
Z11690.1 | - | 1 | 998 |
Set Membership: |
![]() |