siRNA Details
Gene Symbol: Avpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019136.1 | NP_062009.1 | 2 | 433 | ||
XM_006229549.2 | XP_006229611.1 | 3 | 1827 | |||
GenBank mRNA | AY338195.1 | - | 2 | 433 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Avpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019136.1 | NP_062009.1 | 2 | 433 | ||
XM_006229549.2 | XP_006229611.1 | 3 | 1827 | |||
GenBank mRNA | AY338195.1 | - | 2 | 433 |
Set Membership: |
![]() |