siRNA Details
Gene Symbol: Klrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012745.2 | NP_036877.1 | 6 | 807 | ||
XM_006237078.2 | XP_006237140.1 | 6 | 856 | |||
GenBank mRNA | AF009133.1 | - | 6 | 564 | ||
BC086393.1 | - | 6 | 807 | 7303354 |
