siRNA Details
Gene Symbol: Hsd11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017080.2 | NP_058776.2 | 1 | 386 | ||
XM_006250471.2 | XP_006250533.1 | 2 | 167 | |||
XM_006250473.2 | XP_006250535.1 | 2 | 202 | |||
GenBank mRNA | BC078865.1 | - | 1 | 207 | 7109256 | |
CK474613.1 | - | 1 | 403 | |||
FQ209646.1 | - | 1 | 153 | |||
FQ209829.1 | - | 1 | 153 | |||
FQ210302.1 | - | 1 | 151 | |||
FQ210563.1 | - | 1 | 151 | |||
FQ210596.1 | - | 1 | 153 | |||
FQ211059.1 | - | 1 | 150 | |||
FQ211404.1 | - | 1 | 152 | |||
FQ212763.1 | - | 1 | 154 | |||
FQ214152.1 | - | 1 | 153 | |||
FQ214397.1 | - | 2 | 209 | |||
FQ218558.1 | - | 1 | 151 | |||
FQ218584.1 | - | 1 | 153 | |||
FQ219123.1 | - | 1 | 153 | |||
FQ219241.1 | - | 1 | 152 | |||
FQ219385.1 | - | 1 | 151 | |||
FQ219394.1 | - | 1 | 151 | |||
FQ219517.1 | - | 1 | 153 | |||
FQ219624.1 | - | 1 | 86 | |||
J05107.1 | - | 1 | 134 |
