siRNA Details
Gene Symbol: Hsd11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017080.2 | NP_058776.2 | 6 | 1090 | ||
XM_006250471.2 | XP_006250533.1 | 7 | 871 | |||
XM_006250473.2 | XP_006250535.1 | 7 | 906 | |||
GenBank mRNA | AF542063.1 | - | 5 | 690 | ||
AY387051.1 | - | 361 | ||||
BC078865.1 | - | 6 | 911 | 7109256 | ||
FQ209646.1 | - | 6 | 857 | |||
FQ209829.1 | - | 6 | 857 | |||
FQ210302.1 | - | 6 | 855 | |||
FQ210563.1 | - | 6 | 855 | |||
FQ210596.1 | - | 6 | 857 | |||
FQ211059.1 | - | 6 | 854 | |||
FQ211404.1 | - | 6 | 857 | |||
FQ212763.1 | - | 6 | 858 | |||
FQ214152.1 | - | 6 | 857 | |||
FQ214397.1 | - | 7 | 913 | |||
FQ218584.1 | - | 6 | 857 | |||
FQ219123.1 | - | 6 | 857 | |||
FQ219241.1 | - | 6 | 856 | |||
FQ219385.1 | - | 6 | 855 | |||
FQ219394.1 | - | 6 | 856 | |||
FQ219517.1 | - | 6 | 857 | |||
FQ219567.1 | - | 6 | 866 | |||
J05107.1 | - | 6 | 835 |
