siRNA Details
Gene Symbol: Hsd11b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017081.2 | NP_058777.1 | 3 | 690 | ||
XM_006255445.2 | XP_006255507.1 | 3 | 1406 | |||
GenBank mRNA | BC087023.1 | - | 3 | 677 | 7130250 | |
U22424.1 | - | 3 | 691 |
