siRNA Details
Gene Symbol: Itga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030994.2 | NP_112256.1 | 25, 26 | 3583 | ||
XM_008760735.1 | XP_008758957.1 | 24, 25 | 2989 | |||
GenBank mRNA | FQ227807.1 | - | 6, 7 | 548 | ||
X52140.1 | - | 25, 26 | 3607 |
