siRNA Details
Gene Symbol: Aanat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012818.2 | NP_036950.1 | 2 | 254 | ||
XM_006247792.1 | XP_006247854.1 | 2 | 1250 | |||
XM_006247793.1 | XP_006247855.1 | 2 | 1245 | |||
GenBank mRNA | DQ075321.1 | - | 2 | 442 | ||
U29664.1 | - | 1 | 43 | |||
U38306.1 | - | 2 | 254 | |||
U40803.1 | - | 2 | 341 |

More Information
Other Species Targeted

Gene Ontology Categories:
