siRNA Details
Gene Symbol: Stx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012748.1 | NP_036880.1 | 4 | 253 | ||
XM_006249249.2 | XP_006249311.1 | 4 | 411 | |||
XM_006249250.2 | XP_006249312.1 | 4 | 411 | |||
XM_006249253.2 | XP_006249315.1 | 5 | 2157 | |||
XM_008769188.1 | XP_008767410.1 | 5 | 2624 | |||
GenBank mRNA | AY302700.1 | - | 4 | 253 | ||
L20823.1 | - | 4 | 253 | |||
L20888.1 | - | 4 | 253 | |||
L20889.1 | - | 4 | 253 |

More Information
Other Species Targeted

Gene Ontology Categories:
